To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Retinoblastoma Binding Protein 8
- Antigen Symbol:CtIP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:5932
- Antigen Synonyms:Retinoblastoma-interacting protein and myosin-like|CtBP-interacting protein|RBBP-8|CtIP|Sporulation in the absence of SPO11 protein 2 homolog|DNA endonuclease RBBP8|retinoblastoma binding protein 8|Retinoblastoma-binding protein 8|RIMSAE2|EC 3.1
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1.0 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 850 and the C-terminus (residue 897) of human CtBP Interacting Protein (Retinoblastoma Binding Protein 8) using the numbering given in entry NP_002885.1 (GeneID 5932).
- Purification:Immunogen affinity purified
- Cat. No.:102118-148
- Supplier no.:NB100-79810
Specifications
About this item
The CtIP Antibody from Novus Biologicals is a rabbit polyclonal antibody to CtIP. This antibody reacts with human, mouse. The CtIP Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Proximity Ligation Assay.
Type: Primary
Antigen: CtIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse