Order Entry
United States
ContactUsLinkComponent
Anti-CTGF/CCN2 Rabbit Polyclonal Antibody (Biotin)
Anti-CTGF/CCN2 Rabbit Polyclonal Antibody (Biotin)
Catalog # 102115-702
Supplier:  Novus Biologicals
Anti-CTGF/CCN2 Rabbit Polyclonal Antibody (Biotin)
Catalog # 102115-702
Supplier:  Novus Biologicals
Supplier Number:  NB100-724B
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Connective Tissue Growth Factor
  • Antigen Symbol:
    CTGF/CCN2
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    1490
  • Antigen Synonyms:
    Hypertrophic chondrocyte-specific protein 24|Insulin-like growth factor-binding protein 8|HCS24|IBP-8|IGFBP8CCN family member 2|IGF-binding protein 8|MGC102839|NOV2|connective tissue growth factor|CCN2IGFBP-8
  • Storage Buffer:
    PBS
  • Storage Temperature:
    Store at 4C in the dark.
  • Immunogen:
    A synthetic peptide made to a C-terminal portion of human CTGF (between residues 299-349). [UniProt# P29279]
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102115-702
  • Supplier no.:
    NB100-724B

Specifications

About this item

The CTGF / CCN2 Antibody [Biotin] from Novus Biologicals is a rabbit polyclonal antibody to CTGF / CCN2. This antibody reacts with human, mouse, rat. The CTGF / CCN2 Antibody [Biotin] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: CTGF/CCN2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat