To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Connexin 40.1
- Antigen Symbol:Cx40.1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:219770
- Antigen Synonyms:Cx40.1|FLJ90023|Connexin-40.1|connexin 40.1|gap junction delta-4 protein|delta 4|CX40.1connexin40.1|gap junction protein|RP11-425A6.2|40.1kDa
- Storage Buffer:PBS & 2% Sucrose.
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to GJD4(gap junction protein, delta 4, 40.1kDa) The peptide sequence was selected from the middle region of GJD4. Peptide sequence HTKIPDEDESEVTSSASEKLGRQPRGRPHREAAQDPRGSGSEEQPSAAPS.
- Purification:Immunogen affinity purified
- Cat. No.:102192-262
- Supplier no.:NBP1-59280
Specifications
About this item
The Connexin 40.1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Connexin 40.1. This antibody reacts with human. The Connexin 40.1 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: Cx40.1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human