To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CMYA5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:202333
- Antigen Synonyms:myospryn|genethonin-3|C5orf10|cardiomyopathy-associated protein 5|cardiomyopathy associated 5|tripartite motif-containing protein 76|SPRY domain-containing protein 2|dystrobrevin-binding protein 2|DTNBP2|DKFZp451G223|DKFZp451G182|SPRYD2
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human SPRYD2. The immunogen is located within amino acids 3810 - 3860 of SPRYD2.
- Purification:Immunogen affinity purified
- Cat. No.:102210-656
- Supplier no.:NBP1-77117
Specifications
About this item
The CMYA5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CMYA5. This antibody reacts with human, mouse, rat. The CMYA5 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunofluorescence.
Type: Primary
Antigen: CMYA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat