About this item
The CHREBP Antibody from Novus Biologicals is a rabbit polyclonal antibody to CHREBP. This antibody reacts with human, mouse, rat. The CHREBP Antibody has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting.
Type: Primary
Antigen: CHREBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 102131-514
- 102131-528
- Antigen Symbol:
- CHREBP
- CHREBP
- Antigen Name:
- Carbohydrate-responsive element-binding protein
- Carbohydrate-responsive element-binding protein
- Antigen Synonyms:
- Williams Beuren syndrome chromosome region 14|MLX-interacting protein-like|MLX interacting protein-like|Class D basic helix-loop-helix protein 14|MLX interactor|WS-bHLHbHLHd14carbohydrate response element binding protein|CHREBPcarbohydrate-responsive element-binding protein|WBSCR14Williams-Beuren syndrome chromosome region 14 protein 1|BHLHD14|MIOWS basic-helix-loop-helix leucine zipper protein|MONDOBWilliams-Beuren syndrome chromosomal region 14 protein|Williams-Beuren syndrome chromosome region 14 protein 2
- Williams Beuren syndrome chromosome region 14|MLX-interacting protein-like|MLX interacting protein-like|Class D basic helix-loop-helix protein 14|MLX interactor|WS-bHLHbHLHd14carbohydrate response element binding protein|CHREBPcarbohydrate-responsive element-binding protein|WBSCR14Williams-Beuren syndrome chromosome region 14 protein 1|BHLHD14|MIOWS basic-helix-loop-helix leucine zipper protein|MONDOBWilliams-Beuren syndrome chromosomal region 14 protein|Williams-Beuren syndrome chromosome region 14 protein 2
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 51085
- 51085
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- ChIP:
- Yes
- Yes
- Immunogen:
- A C-terminal synthetic peptide made to the human CHREBP protein sequence (between residues 800-852). [UniProt# Q9NP71, Isoform 1/Alpha]
- A C-terminal synthetic peptide made to the human CHREBP protein sequence (between residues 800-852). [UniProt# Q9NP71, Isoform 1/Alpha]
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage Buffer:
- PBS
- PBS
- Storage Temperature:
- Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.