To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:CCDC124
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:115098
- Antigen Synonyms:coiled-coil domain-containing protein 124|coiled-coil domain containing 124
- Storage Buffer:TBS and 0.1% BSA
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:0.2 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 173 and 223 of human coiled-coil domain containing 124 using the numbering given in entry NP_612451.1 (GeneID 115098).
- Purification:Immunogen affinity purified
- Cat. No.:102121-258
- Supplier no.:NB100-97843
The CCDC124 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CCDC124. This antibody reacts with human. The CCDC124 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.
Type: Primary
Antigen: CCDC124
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human