The CARD12 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CARD12. This antibody reacts with human, mouse. The CARD12 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:400
Type: Primary
Antigen: CARD12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 102213-582
- 102213-580
- Antigen Symbol:
- CARD12
- CARD12
- Antigen Name:
- Caspase Recruitment Domain Family, Member 12
- Caspase Recruitment Domain Family, Member 12
- Antigen Synonyms:
- Ipaf|CLAN1ipaf|caspase recruitment domain family|and NACHT-containing protein|CARD domain containing 4|CLR2.1|CARD12leucine rich repeat and CARD domaincontaining 4|member 12|NLR family|Caspase recruitment domain-containing protein 12|Ice protease-activating factor|CLANCARD|LRR|Clan protein
- Ipaf|CLAN1ipaf|caspase recruitment domain family|and NACHT-containing protein|CARD domain containing 4|CLR2.1|CARD12leucine rich repeat and CARD domaincontaining 4|member 12|NLR family|Caspase recruitment domain-containing protein 12|Ice protease-activating factor|CLANCARD|LRR|Clan protein
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 58484
- 58484
- Isotype:
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- A synthetic peptide made to an internal portion of the human CARD12 protein (between residues 600-700) [UniProt Q9NPP4]
- A synthetic peptide made to an internal portion of the human CARD12 protein (between residues 600-700) [UniProt Q9NPP4]
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Concentration:
- 1.13 mg/ml
- 1.13 mg/ml
- Storage Buffer:
- PBS and 30% Glycerol
- PBS and 30% Glycerol
- Storage Temperature:
- Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
- Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.