To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:BRCA1 associated Protein
- Antigen Symbol:BRAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:8315
- Antigen Synonyms:EC 6.3.2.-|BRCA1 associated protein|BRAP2RING finger protein 52|Impedes mitogenic signal propagation|Renal carcinoma antigen NY-REN-63|RNF52galectin-2-binding protein|IMPBRCA1-associated protein
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1.0 mg/ml
- Immunogen:The immunogen for this product maps to a region between residue 560 and 592 of human BRCA1 associated protein using the numbering given in entry NP_006759.3 (GeneID 8315).
- Purification:Immunogen affinity purified
- Cat. No.:102171-504
- Supplier no.:NBP1-42696
The BRAP Antibody from Novus Biologicals is a rabbit polyclonal antibody to BRAP. This antibody reacts with human. The BRAP Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.
Type: Primary
Antigen: BRAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human