To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:betaGal beta-1,3-N-acetylglucosaminyltransferase 5
- Antigen Symbol:B3GNT3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:10331
- Antigen Synonyms:Beta-1,3-Gn-T3|Beta3Gal-T8|TMEM3beta3Gal-T8|UDP-Gal:beta-GlcNAc beta-1,3-galactosyltransferase 8|Beta-1,3-galactosyltransferase 8|HP10328|BGnT-3|B3GN-T3|core1-beta3GlcNAcT|B3GAL-T8|UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase 8|B3GNT-3|Beta-3-Gx-T8|transmembrane protein 3|putative type II membrane protein|B3GALT8|Beta3GalT8|EC 2.4.1.-|EC 2.4.1|beta-1,3-N-acetylglucosaminyltransferase bGnT-3|UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 3|beta3Gn-T3|Beta-1,3-GalTase 8|Core 1 extending beta-1,3-N-acetylglucosaminyltransferase|Beta-1,3-N-acetylglucosaminyltransferase 3
- Storage Buffer:0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human B3GNT3. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Cat. No.:102166-062
- Supplier no.:NBP1-32539
Specifications
About this item
The B3GNT3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to B3GNT3. This antibody reacts with human, mouse. The B3GNT3 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: B3GNT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse