To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Atlastin-2
- Antigen Symbol:ATL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:64225
- Antigen Synonyms:ADP-ribosylation factor-like protein 6-interacting protein 2|ARL3IP2|FLJ23293|aip-2|atlastin-2|ARL6IP2|EC 3.6.5.-|ADP-ribosylation-like factor 6 interacting protein 2|ARL-6-interacting protein 2|atlastin GTPase 2|atlastin2|ADP-ribosylation factor-like 6 interacting protein 2
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:The immunogen for this antibody is Atlastin-2 - C-terminal region. Peptide sequence KQFRSVKKMGGDEFCRRYQDQLEAEIEETYANFIKHNDGKNIFYAARTPA.
- Purification:Immunogen affinity purified
- Cat. No.:102223-104
- Supplier no.:NBP1-98394
Specifications
About this item
The Atlastin-2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Atlastin-2. This antibody reacts with mouse. The Atlastin-2 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: Atlastin-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse