To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:AT Rich Interactive Domain 2
- Antigen Symbol:ARID2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:196528
- Antigen Synonyms:BRG1-associated factor 200|DKFZp686G052|ARID domain-containing protein 2|p200|FLJ30619|DKFZp779P0222|zipzap|zipzap/p200|BAF200KIAA1557AT-rich interactive domain-containing protein 2|AT rich interactive domain 2 (ARID|Zinc finger protein with activation potential|RFX-like)
- Storage Buffer:Tris-Citrate/Phosphate (pH 7.0 - 8.0)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:1.0 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between residue 1600 and 1650 of human AT rich interactive domain 2 using the numbering given in entry NP_689854.2 (GeneID 196528).
- Purification:Immunogen affinity purified
- Cat. No.:102161-084
- Supplier no.:NBP1-26614
Specifications
About this item
The ARID2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ARID2. This antibody reacts with human. The ARID2 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.
Type: Primary
Antigen: ARID2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human