Order Entry
United States
ContactUsLinkComponent
Anti-Archaemetzincin 2 Rabbit Polyclonal Antibody
Anti-Archaemetzincin 2 Rabbit Polyclonal Antibody
Catalog # 102214-890
Supplier:  Novus Biologicals
Anti-Archaemetzincin 2 Rabbit Polyclonal Antibody
Catalog # 102214-890
Supplier:  Novus Biologicals
Supplier Number:  NBP1-79621
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Archaemetzincin 2
  • Antigen Symbol:
    AMZ2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    51321
  • Antigen Synonyms:
    EC 3.-|Archeobacterial metalloproteinase-like protein 2|archaemetzincins-2|archaelysin family metallopeptidase 2|archaemetzincin-2
  • Storage Buffer:
    PBS and 2% Sucrose
  • Storage Temperature:
    Store at -20C. Avoid freeze-thaw cycles.
  • Immunogen:
    Synthetic peptide directed towards the C terminal of human AMZ2. Peptide sequence ACLMQGSNHLEEADRRPLNLCPICLHKLQCAVGFSIVERYKALVRWIDDE.
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    102214-890
  • Supplier no.:
    NBP1-79621

Specifications

About this item

The Archaemetzincin 2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Archaemetzincin 2. This antibody reacts with human. The Archaemetzincin 2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Archaemetzincin 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human