To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Apolipoprotein B mRNA Editing Enzyme, Catalytic Polypeptide 1
- Antigen Symbol:APOBEC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:339
- Antigen Synonyms:Apolipoprotein B mRNA-editing enzyme 1|CDAR1|catalytic polypeptide 1|U-editing enzyme APOBEC-1|EC 3.5.4.-|EC 3.5.4|apolipoprotein B mRNA editing enzyme|HEPRapolipoprotein B mRNA editing enzyme complex-1|APOBEC-1|BEDPC->
- Storage Buffer:PBS & 2% Sucrose.
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to APOBEC1(apolipoprotein B mRNA editing enzyme, catalytic polypeptide 1) The peptide sequence was selected from the N terminal of APOBEC1. Peptide sequence TSEKGPSTGDPTLRRRIEPWEFDVFYDPRELRKEACLLYEIKWGMSRKI
- Purification:Immunogen affinity purified
- Cat. No.:102189-432
- Supplier no.:NBP1-57268
Specifications
About this item
The APOBEC1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to APOBEC1. This antibody reacts with human. The APOBEC1 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: APOBEC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human