About this item
The AG-2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to AG-2. This antibody reacts with human. The AG-2 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100
Type: Primary
Antigen: AG-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 102121-586
- 102121-588
- Antigen Symbol:
- AG-2
- AG-2
- Antigen Name:
- Anterior Gradient 2
- Anterior Gradient 2
- Antigen Synonyms:
- AG2member 17|secreted cement gland homolog|hAG-2|AG-2|anterior gradient 2 homolog (Xenopus laevis)|anterior gradient homolog 2 (Xenopus laevis)|HPC8|GOB-4
- AG2member 17|secreted cement gland homolog|hAG-2|AG-2|anterior gradient 2 homolog (Xenopus laevis)|anterior gradient homolog 2 (Xenopus laevis)|HPC8|GOB-4
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 10551
- 10551
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- A synthetic peptide corresponding to an internal region of human AGR2 (within residues 50-100). [UniProt# O95994]
- A synthetic peptide corresponding to an internal region of human AGR2 (within residues 50-100). [UniProt# O95994]
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Concentration:
- 1.0 mg/ml
- 1.0 mg/ml
- Storage Buffer:
- PBS and 30% Glycerol
- PBS and 30% Glycerol
- Storage Temperature:
- Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.