To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:ADAM Metallopeptidase With Thrombospondin Type 1 Motif, 6
- Antigen Symbol:ADAMTS6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:11174
- Antigen Synonyms:ADAMTS-6|ADAM-TS 6|EC 3.4.24.82|ADAM metallopeptidase with thrombospondin type 1 motif|66,EC 3.4.24.-
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptide corresponding to a.a. 563-897 of Human ADAMTS6 (NP_922932). Peptide Sequence: PWSECSATCAGGVQRQEVVCKRLDDNSIVQNNYCDPDSKPPENQRACNTE
- Purification:Immunogen affinity purified
- Cat. No.:102215-402
- Supplier no.:NBP1-79877
The ADAMTS6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ADAMTS6. This antibody reacts with human. The ADAMTS6 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: ADAMTS6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human