Anti-ACTR-IC Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The ALK-7 / Activin Receptor Type 1C Antibody from Novus Biologicals is a rabbit polyclonal antibody to ALK-7 / Activin Receptor Type 1C. This antibody reacts with human, mouse, rat. The ALK-7 / Activin Receptor Type 1C Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: ACTR-IC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
NBP1-50659.pdf
Datasheet
Specifications
- Size:100 μg
- Clonality:Polyclonal
- Purification:Immunogen affinity purified
- Storage Buffer:PBS (pH 7.2), 0.5% BSA and 30% Glycerol
- Isotype:IgG
- Host:Rabbit
- Concentration:0.5 mg/ml
- Cat. No.:102180-456
- Reactivity:Human|Rat|Mouse
- Western Blot:Yes
- Antibody Type:Primary
- Antigen Symbol:ACTR-IC
- Gene ID:130399
- Immunogen:Synthetic peptide surrounding amino acid 155 of mouse Activin Receptor Type 1C
- Conjugation:Unconjugated
- Antigen Synonyms:activin A receptor|ACVRLK7|ALK7ALK-7|type IC|Activin receptor-like kinase 7|ACTR-IC|EC 2.7.11.30|EC 2.7.11|activin receptor type-1C|Activin receptor type IC
- Antigen Name:Activin Receptor Type 1C
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



