To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:ACF
- Antigen Symbol:ACF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:29974
- Antigen Synonyms:APOBEC-1 stimulating protein|apo-B RNA editing protein|APOBEC1-stimulating protein|APOBEC1 complementation factor|ACF65|ACFASPACF64|APOBEC1CF|MGC163391|apobec-1 complementation factor (ACF) (ASP)
- Storage Buffer:PBS & 2% Sucrose.
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptides corresponding to A1CF(APOBEC1 complementation factor) The peptide sequence was selected from the N terminal of A1CF. Peptide sequence EAVCLGTCPEPEASMSTAIPGLKKGNNALQSIILQTLLEKENGQRKYGGP.
- Purification:Immunogen affinity purified
- Cat. No.:102189-510
- Supplier no.:NBP1-57308
Specifications
About this item
The ACF Antibody from Novus Biologicals is a rabbit polyclonal antibody to ACF. This antibody reacts with human. The ACF Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: ACF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human