To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Acetyl-CoA Acetyltransferase 1
- Antigen Symbol:ACAT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:6646
- Antigen Synonyms:ACACT|ACATACAT1|Acyl-coenzyme A:cholesterol acyltransferase 1|RP11-215I23.2|sterol O-acyltransferase 1|EC 2.3.1.26|acyl-Coenzyme A: cholesterol acyltransferase|ACACT1|ACAT-1|Cholesterol acyltransferase 1|sterol O-acyltransferase (acyl-Coenzyme A: cholesterol acyltransferase) 1|STATSOAT
- Storage Buffer:PBS and 2% Sucrose
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptide directed towards the middle region of human SOAT1 (NP_003092). Peptide sequence ASRFIIIFEQIRFVMKAHSFVRENVPRVLNSAKEKSSTVPIPTVNQYLYF.
- Purification:Immunogen affinity purified
- Cat. No.:102216-740
- Supplier no.:NBP1-80547
Specifications
About this item
The ACAT Antibody from Novus Biologicals is a rabbit polyclonal antibody to ACAT. This antibody reacts with human. The ACAT Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: ACAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human