To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:5-Hydroxytryptamine receptor 7
- Antigen Symbol:5-HT7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:3363
- Antigen Synonyms:5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled)|5-HT75-hydroxytryptamine receptor 7|Serotonin receptor 7|5-HT-7|serotonin 5-HT-7 receptor|5-HT-X
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:This antibody was developed by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 13-28 of the rat 5-HT7R (AAA42134.1).
- Purification:Protein G purified
- Cat. No.:102109-922
- Supplier no.:NB100-56352
Specifications
About this item
The 5-HT7 Antibody from Novus Biologicals is a rabbit polyclonal antibody to 5-HT7. This antibody reacts with human, mouse, rat, canine. The 5-HT7 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: 5-HT7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Canine