Order Entry
United States
ContactUsLinkComponent
Pepsin
Pepsin
Catalog # 102136-948
Supplier:  Novus Biologicals
CAS Number:  
Pepsin
Catalog # 102136-948
Supplier:  Novus Biologicals
Supplier Number:  NB900-66724
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Source:
  • Species:
  • Size:
    50 ml
  • Environmentally Preferable:
  • Storage Conditions:
    Store at 4 °C. Do not freeze.
  • Enzyme Name:
    Pepsin
  • Cat. No.:
    102136-948
  • Supplier no.:
    NB900-66724

Specifications

About this item

Stablized Pepsin solution for antigen retrieval

  • No Preservative

Dilutions: Immunohistochemistry 1:10-1:500
Application Notes: 1. Deparaffinize the tissue sections and bring them to buffer. 2. Remove excess buffer from the glass without letting the tissue dry. 3. Depending on the size of the tissue section, add 1 or 2 drops of pepsin solution. 4. Incubate the slide for 5 to 10 minutes at 37C. 5. Wash slides with buffer and proceed for the immunostaining.