About this item
The Vimentin Antibody (2A52) from Novus Biologicals is a mouse monoclonal antibody to Vimentin. This antibody reacts with human, mouse, rat, porcine, bovine. The Vimentin Antibody (2A52) has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: Vimentin
Clonality: Monoclonal
Clone: 2A52
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Porcine, Bovine
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 102217-898
- 102217-900
- Antigen Symbol:
- VIM
- VIM
- Antigen Name:
- Vimentin
- Vimentin
- Antigen Synonyms:
- vimentin|FLJ36605
- vimentin|FLJ36605
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 2A52
- 2A52
- Reactivity:
- Pig, Human, Bovine, Rat, Mouse
- Pig, Human, Bovine, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 7431
- 7431
- Isotype:
- IgG1
- IgG1
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant human Vimentin expressed in and purified from E. coli. [UniProt# P08670]
- Recombinant human Vimentin expressed in and purified from E. coli. [UniProt# P08670]
- Purification:
- Protein G purified
- Protein G purified
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage Buffer:
- PBS
- PBS
- Storage Temperature:
- Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.