To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:TNF Receptor-associated Factor 2
- Antigen Symbol:TRAF-2
- Clonality:Monoclonal
- Clone:33A1293
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:7186
- Antigen Synonyms:EC 6.3.2.-|MGC:45012|Tumor necrosis factor type 2 receptor-associated protein 3|TRAP3TRAP|tumor necrosis factor type 2 receptor associated protein 3|TNF receptor-associated factor 2|E3 ubiquitin-protein ligase TRAF2
- Storage Buffer:PBS containing 0.05% BSA
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Anti-TRAF2 monoclonal antibody was raised against a fusion protein corresponding to amino acids 205 to 222 of human TRAF2.
- Purification:Protein G purified
- Cat. No.:102110-258
- Supplier no.:NB100-56715
The TRAF-2 Antibody (33A1293) from Novus Biologicals is a mouse monoclonal antibody to TRAF-2. This antibody reacts with human. The TRAF-2 Antibody (33A1293) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: TRAF-2
Clonality: Monoclonal
Clone: 33A1293
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human