To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:TCF-3/E2A
- Clonality:Monoclonal
- Clone:6B8
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:6929
- Antigen Synonyms:transcription factor 3 (E2A immunoglobulin enhancer binding factors E12/E47)|VDIR|transcription factor E2-alpha|Immunoglobulin enhancer-binding factor E12/E47|MGC129647|E2A immunoglobulin enhancer-binding factor E12/E47|ITF1Transcription factor ITF-1|bHLHb21TCF-3|MGC129648|Transcription factor 3|VDR interacting repressor|E2AKappa-E2-binding factor|Class B basic helix-loop-helix protein 21|Immunoglobulin transcription factor 1|BHLHB21
- Storage Buffer:Ascites
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Purified recombinant fragment of human TCF3 expressed in E. Coli.
- Purification:Unpurified
- Cat. No.:102171-248
- Supplier no.:NBP1-42566
Specifications
About this item
The TCF-3 / E2A Antibody (6B8) from Novus Biologicals is a mouse monoclonal antibody to TCF-3 / E2A. This antibody reacts with human. The TCF-3 / E2A Antibody (6B8) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA.
Type: Primary
Antigen: TCF-3/E2A
Clonality: Monoclonal
Clone: 6B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human