To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:SIL1 Nucleotide Exchange Factor
- Antigen Symbol:SIL1
- Clonality:Monoclonal
- Clone:3000
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:64374
- Antigen Synonyms:ULG5|SIL1 homolog|MSS|endoplasmic reticulum chaperone (S. cerevisiae)|nucleotide exchange factor SIL1|BAPMarinesco-Sjogren syndrome|BiP-associated protein
- Storage Buffer:PBS (pH 7.3), 1.0% BSA and 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human SIL1 (NP_071909) produced in HEK293T cell.
- Purification:Immunogen affinity purified
- Cat. No.:102231-490
- Supplier no.:NBP2-02377
Specifications
About this item
The SIL1 Antibody (3E3) from Novus Biologicals is a mouse monoclonal antibody to SIL1. This antibody reacts with human. The SIL1 Antibody (3E3) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: SIL1
Clonality: Monoclonal
Clone: 3000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human