To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Serpin Family B Member 13
- Antigen Symbol:SERPINB13
- Clonality:Monoclonal
- Clone:2B6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:5275
- Antigen Synonyms:HUR 7|Peptidase inhibitor 13|HURPIN|HaCaT UV-repressible serpin|serpin peptidase inhibitor|Headpin|UV-B repressed sequence|serpin B13|Proteinase inhibitor 13|serine (or cysteine) proteinase inhibitor|PI13hurpin|protease inhibitor 13 (hurpin|clade B (ovalbumin)|HUR7|headpin)|HSHUR7SEQ|MGC126870|member 13|PI-13
- Storage Buffer:PBS (pH 7.3), 1.0% BSA and 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human SERPINB13(NP_036529) produced in HEK293T cell.
- Purification:Immunogen affinity purified
- Cat. No.:102229-360
- Supplier no.:NBP2-01312
Specifications
About this item
The SERPINB13 Antibody (2B6) from Novus Biologicals is a mouse monoclonal antibody to SERPINB13. This antibody reacts with human. The SERPINB13 Antibody (2B6) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: SERPINB13
Clonality: Monoclonal
Clone: 2B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human