To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Regulator of G-protein Signaling 16
- Antigen Symbol:RGS16
- Clonality:Monoclonal
- Clone:4E5
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:6004
- Antigen Synonyms:RGSR|A28-RGS14P|hRGS-r|regulator of G-protein signaling 16|regulator of G-protein signalling 16|Retinal-specific RGS|A28-RGS14|RGS-r|Retinally abundant regulator of G-protein signaling
- Storage Buffer:PBS (pH 7.3), 1.0% BSA and 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human RGS16(NP_002919) produced in HEK293T cell.
- Purification:Immunogen affinity purified
- Cat. No.:102229-904
- Supplier no.:NBP2-01584
Specifications
About this item
The RGS16 Antibody (4E5) from Novus Biologicals is a mouse monoclonal antibody to RGS16. This antibody reacts with human, mouse. The RGS16 Antibody (4E5) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: RGS16
Clonality: Monoclonal
Clone: 4E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse