To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Repulsive Guidance Molecule C
- Antigen Symbol:RGM-C
- Clonality:Monoclonal
- Clone:Fery-1
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Size:0.05 mg
- Environmentally Preferable:
- Gene ID:148738
- Antigen Synonyms:repulsive guidance molecule c|hemochromatosis type 2 (juvenile)|hemojuvelin|HFE2AMGC23953|RGMCRGM domain family member C|HJVHemochromatosis type 2 protein|haemojuvelin|JH
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Recombinant human RGMC (aa 36-398).
- Purification:Protein G purified
- Cat. No.:102222-150
- Supplier no.:NBP1-97611
Specifications
About this item
The RGM-C / Hemojuvelin Antibody (Fery-1) from Novus Biologicals is a mouse monoclonal antibody to RGM-C / Hemojuvelin. This antibody reacts with human. The RGM-C / Hemojuvelin Antibody (Fery-1) has been validated for the following applications: Flow Cytometry, Immunoprecipitation.
Type: Primary
Antigen: RGM-C
Clonality: Monoclonal
Clone: Fery-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human