To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Folate Hydrolase 1
- Antigen Symbol:PSMA/FOLH1/NAALADase I
- Clonality:Monoclonal
- Clone:3H5
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Monkey,Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:2346
- Antigen Synonyms:glutamate carboxylase II|EC 3.4.17.21|GCPIImGCP|folate hydrolase (prostate-specific membrane antigen) 1|Prostate-specific membrane antigen|Folylpoly-gamma-glutamate carboxypeptidase|Membrane glutamate carboxypeptidase|GCP2|Cell growth-inhibiting gene 27 protein|PSMPteroylpoly-gamma-glutamate carboxypeptidase|Glutamate carboxypeptidase II|FOLHNAALADase I|NAALAdase|PSMAFGCP|N-acetylated alpha-linked acidic dipeptidase 1|NAALAD1N-acetylated-alpha-linked acidic dipeptidase I|cell growth-inhibiting protein 27|Folate hydrolase 1|prostate specific membrane antigen variant F|glutamate carboxypeptidase 2
- Storage Buffer:PBS (pH 7.3), 1.0% BSA and 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human FOLH1(NP_004467) produced in HEK293T cell.
- Purification:Immunogen affinity purified
- Cat. No.:102230-826
- Supplier no.:NBP2-02045
Specifications
About this item
The PSMA / FOLH1 / NAALADase I Antibody (3H5) from Novus Biologicals is a mouse monoclonal antibody to PSMA / FOLH1 / NAALADase I. This antibody reacts with human, mouse, rat, monkey. The PSMA / FOLH1 / NAALADase I Antibody (3H5) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: PSMA/FOLH1/NAALADase I
Clonality: Monoclonal
Clone: 3H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Monkey