To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Proteasome 20S alpha 6
- Antigen Symbol:PSMA6
- Clonality:Monoclonal
- Clone:4C9
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Primates,Human,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:5687
- Antigen Synonyms:macropain) subunit|6|PROS2727 kDa prosomal protein|MGC2333|p27KPROS-27|Macropain iota chain|EC 3.4.25.1|alpha type|P27K|proteasome subunit iota|proteasome (prosome|Proteasome iota chain|macropain subunit iota|proteasome subunit alpha type-6|MGC23846|Multicatalytic endopeptidase complex iota chain|IOTA|MGC22756|prosomal P27K protein
- Storage Buffer:PBS (pH 7.3), 1.0% BSA and 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human PSMA6 (NP_002782) produced in E.coli.
- Purification:Immunogen affinity purified
- Cat. No.:102234-408
- Supplier no.:NBP2-03836
Specifications
About this item
The Proteasome 20S alpha 6 Antibody (4C9) from Novus Biologicals is a mouse monoclonal antibody to Proteasome 20S alpha 6. This antibody reacts with human, mouse, rat, canine, primate. The Proteasome 20S alpha 6 Antibody (4C9) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Proteasome 20S alpha 6
Clonality: Monoclonal
Clone: 4C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat, Canine, Primate