To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Presenilin 2
- Antigen Symbol:Presenilin-2
- Clonality:Monoclonal
- Clone:APS 26
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Primates,Human,Rat,Mouse
- Western Blot:Yes
- Size:200ug
- Environmentally Preferable:
- Epitope:Endoplasmic reticulum and Golgi Apparatus: endoplasmic reticulum membrane - multi-pass membrane protein; Golgi membrane - multi-pass membrane protein.
- Gene ID:5664
- Antigen Synonyms:STM2STM-2|E5-1|AD3LP|PS-2|EC 3.4.23|PSNL2|EC 3.4.23.-|PS2CMD1V|AD4AD3L|presenilin 2 (Alzheimer disease 4)|presenilin-2|AD5
- Storage Buffer:PBS with 1mg/ml BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide corresponding to residues C L(317) P Y D P E M E E D S Y D S F G E P(334) of human PS2.
- Purification:Protein G purified
- Cat. No.:102116-442
- Supplier no.:NB100-74513
Specifications
About this item
The Presenilin-2 Antibody (APS 26) from Novus Biologicals is a mouse monoclonal antibody to Presenilin-2. This antibody reacts with human, mouse, rat, primate. The Presenilin-2 Antibody (APS 26) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Presenilin-2
Clonality: Monoclonal
Clone: APS 26
Conjugation: Unconjugated
Epitope: Endoplasmic reticulum and Golgi Apparatus: endoplasmic reticulum membrane - multi-pass membrane protein; Golgi membrane - multi-pass membrane protein.
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Primate