To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Penicillin
- Antigen Symbol:PCN
- Clonality:Monoclonal
- Clone:P2B9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:
- Size:100ug
- Environmentally Preferable:
- Antigen Synonyms:Phenoxymethylpenicillin|flucloxacillin|dicloxacillin|methicillin|Penicilin|Ampicillin|Benzylpenicillin|PCN|Penicillin G
- Storage Buffer:10mM Potassium Phosphate, 2.5mM Potassium Chloride, 135mM NaCl and 1.0 mg/ml BSA
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Penicillin-BSA
- Tested Applications:, Radioimmunoassay
- Purification:Protein A purified
- Cat. No.:102212-970
- Supplier no.:NBP1-78588
Specifications
About this item
The Penicillin Antibody (P2B9) from Novus Biologicals is a mouse monoclonal antibody to Penicillin. This antibody reacts with other. The Penicillin Antibody (P2B9) has been validated for the following applications: ELISA, Immunoprecipitation, Radioimmunoassay.
Type: Primary
Antigen: Penicillin
Clonality: Monoclonal
Clone: P2B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Other