To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:pan Cadherin
- Antigen Symbol:pan Cadherin
- Clonality:Monoclonal
- Clone:CH-19
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Rat,Mouse,Cat,Hamster,Pig,Rabbit,Xenopus,Sheep,Goat,Chicken,Human,Bovine,Guinea pig,Dog
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:999
- Antigen Synonyms:cadherin-1|CD324|cadherin 1|ECAD|CD324 antigen|Epithelial cadherin|type 1|Arc-1|CAM 120/80|cell-CAM 120/80|UVOE-Cadherin|uvomorulin|E-cadherin (epithelial)|calcium-dependent adhesion protein|epithelial|LCAM|E-cadherin|CDHE
- Storage Buffer:Ascites
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Synthetic peptide corresponding to the C-terminal amino acids of chicken N-Cadherin with an extra N-terminal lysine residue (24 amino acids) coupled to KLH.
- Tested Applications:Microarray
- Purification:Unpurified
- Cat. No.:102106-322
- Supplier no.:NB100-1907
The pan Cadherin Antibody (CH-19) from Novus Biologicals is a mouse monoclonal antibody to pan Cadherin. This antibody reacts with human, mouse, rat, porcine, bovine, canine, chicken, feline, guinea pig, goat, hamster, rabbit, sheep, xenopus. The pan Cadherin Antibody (CH-19) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Microarray.
Recommended Dilutions: Immunohistochemistry (Parafin): 1/500
Type: Primary
Antigen: pan Cadherin
Clonality: Monoclonal
Clone: CH-19
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Feline, Guinea Pig, Goat, Hamster, Rabbit, Sheep, Xenopus