To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cyclin-dependent kinase inhibitor 1C
- Antigen Symbol:p57 Kip2
- Clonality:Monoclonal
- Clone:3000
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:1028
- Antigen Synonyms:Kip2)|KIP2BWCR|cyclin-dependent kinase inhibitor 1C|p57|WBS|cyclin-dependent kinase inhibitor 1C (p57|Cyclin-dependent kinase inhibitor p57|p57Kip2|Beckwith-Wiedemann syndrome|BWS
- Storage Buffer:Ascites
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Purified recombinant fragment of human CDKN1C expressed in E. Coli.
- Purification:Unpurified
- Cat. No.:102182-066
- Supplier no.:NBP1-51675
Specifications
About this item
The p57 Kip2 Antibody (3E3) from Novus Biologicals is a mouse monoclonal antibody to p57 Kip2. This antibody reacts with human. The p57 Kip2 Antibody (3E3) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: p57 Kip2
Clonality: Monoclonal
Clone: 3000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human