To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:KIR2.3
- Clonality:Monoclonal
- Clone:S25-35
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:3761
- Antigen Synonyms:subfamily J|hippocampal inward rectifier potassium channel|Kir2.3|IRK-3|inwardly rectifying subfamily J member 4|Potassium channel|member 4|HIRhIRK2|HRK1MGC142066|Hippocampal inward rectifier|inward rectifier potassium channel 4|Inward rectifier K(+) channel Kir2.3|inward rectifier K+ channel Kir2.3|potassium inwardly-rectifying channel|HIRK2|IRK3MGC142068
- Storage Buffer:PBS (pH 7.4) and 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Fusion protein amino acids 390-445 of human Kir2.3
- Purification:Protein G purified
- Cat. No.:102252-600
- Supplier no.:NBP2-12902
Specifications
About this item
Mouse Monoclonal KIR2.3 Antibody (S25-35). Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat.
Recommended Dilutions: Immunohistochemistry (Parafin): 0.1-1.0 ?g/mL
Type: Primary
Antigen: KIR2.3
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human