Order Entry
United States
ContactUsLinkComponent
Anti-FABP2/I-FABP Mouse Monoclonal Antibody [clone: 9A9B7B3]
Anti-FABP2/I-FABP Mouse Monoclonal Antibody [clone: 9A9B7B3]
Catalog # 102181-898
Supplier:  Novus Biologicals
Anti-FABP2/I-FABP Mouse Monoclonal Antibody [clone: 9A9B7B3]
Catalog # 102181-898
Supplier:  Novus Biologicals
Supplier Number:  NBP1-51589
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    intestinal FABP
  • Antigen Symbol:
    FABP2/I-FABP
  • Clonality:
    Monoclonal
  • Clone:
    9A9B7B3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    2169
  • Antigen Synonyms:
    I-FABPMGC133132|Intestinal-type fatty acid-binding protein|fatty acid-binding protein|Fatty acid-binding protein 2|fatty acid binding protein 2|intestinal|FABPI
  • Storage Buffer:
    Ascites
  • Storage Temperature:
    Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
  • Immunogen:
    Purified recombinant fragment of human FABP2 expressed in E. Coli.
  • Purification:
    Unpurified
  • Cat. No.:
    102181-898
  • Supplier no.:
    NBP1-51589

Specifications

About this item

The FABP2 / I-FABP Antibody (9A9B7B3) from Novus Biologicals is a mouse monoclonal antibody to FABP2 / I-FABP. This antibody reacts with human, mouse. The FABP2 / I-FABP Antibody (9A9B7B3) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: FABP2/I-FABP
Clonality: Monoclonal
Clone: 9A9B7B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse