About this item
Mouse Monoclonal HIP1 Related Antibody (1E1). Tested Applications: Western Blot. Tested Reactivity: Human, Rat.
Type: Primary
Antigen: HIP1 Related
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 102130-216
- 102130-214
- Antigen Symbol:
- HIP1 Related
- HIP1 Related
- Antigen Name:
- HIP1 Related
- HIP1 Related
- Antigen Synonyms:
- Huntingtin-interacting protein 12|huntingtin interacting protein 1 related|huntingtin interacting protein 12|FLJ14000|ILWEQ|HIP12HIP1-related protein|MGC47513|HIP-12|KIAA0655FLJ27022|huntingtin-interacting protein 1-related protein|HIP3
- Huntingtin-interacting protein 12|huntingtin interacting protein 1 related|huntingtin interacting protein 12|FLJ14000|ILWEQ|HIP12HIP1-related protein|MGC47513|HIP-12|KIAA0655FLJ27022|huntingtin-interacting protein 1-related protein|HIP3
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 10
- 10
- Reactivity:
- Bacteria, Human, Rat, Mouse
- Bacteria, Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 9026
- 9026
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Immunogen:
- Human HIP1 Related-TALIN Homology Region (57 kDa). [UniProt# O75146]
- Human HIP1 Related-TALIN Homology Region (57 kDa). [UniProt# O75146]
- Purification:
- Unpurified
- Unpurified
- Storage Buffer:
- Ascites
- Ascites
- Storage Temperature:
- Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.