To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Endorepellin/Perlecan/Heparan Sulfate Proteoglycan
- Clonality:Monoclonal
- Clone:A76
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Bovine
- Size:100 ug
- Environmentally Preferable:
- Gene ID:3339
- Antigen Synonyms:endorepellin (domain V region)|SJS|HSPG|heparan sulfate proteoglycan 2|PLCSchwartz-Jampel syndrome 1 (chondrodystrophic myotonia)|basement membrane-specific heparan sulfate proteoglycan core protein|SJA|perlecan proteoglycan|PRCAN|perlecan|SJS1
- Storage Buffer:10mM Phosphate (pH 7.4) and 0.5M NaCl
- Storage Temperature:Store at 4C in the dark.
- Concentration:1.0 mg/ml
- Immunogen:Lysed bovine corneal endothelial cells and extracellular matrix
- Purification:Protein A or G purified
- Cat. No.:102156-640
- Supplier no.:NBP1-05170
Specifications
About this item
The Endorepellin / Perlecan / Heparan Sulfate Proteoglycan Antibody (A76) - BSA Free from Novus Biologicals is a mouse monoclonal antibody to Endorepellin / Perlecan / Heparan Sulfate Proteoglycan. This antibody reacts with human, bovine. The Endorepellin / Perlecan / Heparan Sulfate Proteoglycan Antibody (A76) - BSA Free has been validated for the following applications: ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Type: Primary
Antigen: Endorepellin/Perlecan/Heparan Sulfate Proteoglycan
Clonality: Monoclonal
Clone: A76
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Bovine