To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:HAP1
- Clonality:Monoclonal
- Clone:1B6
- Conjugation:Biotin
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:9001
- Antigen Synonyms:HLPHAP-1|Neuroan 1|HLP1|huntingtin-associated protein 1|HAP2huntingtin-associated protein 2|HIP5|hHLP1
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Fusion protein from rat HAP1. [UniProt# P54256]
- Purification:Protein G purified
- Cat. No.:102124-782
- Supplier no.:NB110-74569B
Mouse Monoclonal HAP1 Antibody (1B6) [Biotin]. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin, Immunoprecipitation. Tested Reactivity: Human, Mouse, Rat.
Type: Primary
Antigen: HAP1
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human