To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:GM-CSF R alpha
- Clonality:Monoclonal
- Clone:MM0316-8W1
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Form:Lyophilized
- Gene ID:1438
- Antigen Synonyms:CD116 antigen|alpha|GM-CSF receptor alpha subunit|MGC4838|GMCSFR|colony stimulating factor 2 receptor alpha subunit|GMCSFR-alpha|granulocyte-macrophage colony-stimulating factor receptor subunit alpha|GMR-alpha|CSF2RAX|CSF2RY|CSF2RAY|GMR|colony stimulating factor 2 receptor|granulocyte-macrophage colony-stimulating factor receptor alpha chain|GM-CSF-R-alpha|CDw116|MGC3848|low-affinity(granulocyte-macrophage)|CSF2R|CD116|CSF2RX
- Storage Buffer:A 0.2 um filtered solution in PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Human GM-CSF Receptor alpha
- Purification:Protein G purified
- Cat. No.:102251-420
- Supplier no.:NBP2-12342
Specifications
About this item
Mouse Monoclonal GM-CSF Receptor alpha Antibody (MM0316-8W1). Tested Applications: Western Blot, Flow Cytometry. Tested Reactivity: Human.
Type: Primary
Antigen: GM-CSF Receptor alpha
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human