To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:FGF R2
- Clonality:Monoclonal
- Clone:5C5
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:2263
- Antigen Synonyms:TK14|CD332 antigen|ECT1|EC 2.7.10|TK25|BEK|soluble FGFR4 variant 4|K-sam|JWS|KGFR|CFD1|EC 2.7.10.1|craniofacial dysostosis 1|Keratinocyte growth factor receptorreceptor like 14|FLJ98662|CEK3|CD332|fibroblast growth factor receptor 2|KSAM|BBDS|BFR-1|Jackson-Weiss syndrome
- Storage Buffer:PBS (pH 7.3), 1.0% BSA and 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human FGFR2 (NP_000132) produced in HEK293T cell.
- Purification:Immunogen affinity purified
- Cat. No.:102228-604
- Supplier no.:NBP2-00932
Specifications
About this item
The FGF R2 Antibody (5C5) from Novus Biologicals is a mouse monoclonal antibody to FGF R2. This antibody reacts with human. The FGF R2 Antibody (5C5) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: FGF R2
Clonality: Monoclonal
Clone: 5C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human