To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Chemokine (C-X-C Motif) Receptor 2
- Antigen Symbol:CXCR2
- Clonality:Monoclonal
- Clone:48311.211
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Size:100 μg
- Environmentally Preferable:
- Neutralisation:Yes
- Gene ID:3579
- Antigen Synonyms:CXCR2 gene for IL8 receptor type B|IL-8 receptor type 2|IL-8R B|CXC-R2|chemokine (C-X-C motif) receptor 2|CD182 antigen|IL8R2|CDw128b|GRO/MGSA receptor|chemokine (CXC) receptor 2|interleukin 8 receptor B|High affinity interleukin-8 receptor B|IL8RA|C-X-C chemokine receptor type 2|interleukin 8 receptor type 2|CXCR-2|beta|interleukin-8 receptor type B
- Storage Buffer:10mM Sodium Phosphate (pH 7.6), 0.25M NaCl and 15 mg/ml BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Human CXCR2 transfected NSO mouse myeloma cells.
- Purification:Protein G purified
- Cat. No.:102105-676
- Supplier no.:NB100-1631
Specifications
About this item
The CXCR2 / IL-8 RB Antibody (48311.211) from Novus Biologicals is a mouse monoclonal antibody to CXCR2 / IL-8 RB. This antibody reacts with human. The CXCR2 / IL-8 RB Antibody (48311.211) has been validated for the following applications: Flow Cytometry, Blocking / Neutralizing.
Type: Primary
Antigen: CXCR2
Clonality: Monoclonal
Clone: 48311.211
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human