To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CD3 epsilon
- Antigen Symbol:CD3 epsilon
- Clonality:Monoclonal
- Clone:TR66
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 μg
- Environmentally Preferable:
- Gene ID:916
- Antigen Synonyms:CD3e molecule|epsilon (CD3-TCR complex)|CD3e antigen|TCRE|epsilon subunit of T3|T3E|FLJ18683|CD3-epsilon|T-cell surface glycoprotein CD3 epsilon chain|epsilon polypeptide (TiT3 complex)|T-cell antigen receptor complex|T-cell surface antigen T3/Leu-4 epsilon chain
- Storage Buffer:PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Cloned human helper T cell line.
- Purification:Protein G purified
- Cat. No.:102221-840
- Supplier no.:NBP1-97446
Specifications
About this item
The CD3 epsilon Antibody (TR66) from Novus Biologicals is a mouse monoclonal antibody to CD3 epsilon. This antibody reacts with human. The CD3 epsilon Antibody (TR66) has been validated for the following applications: Flow Cytometry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation.
Type: Primary
Antigen: CD3 epsilon
Clonality: Monoclonal
Clone: TR66
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human