To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:CD1
- Clonality:Monoclonal
- Clone:76-7-4
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Pig
- Size:0.25 mg
- Environmentally Preferable:
- Gene ID:909
- Antigen Synonyms:FCB6|epidermal dendritic cell marker CD1a|cortical thymocyte antigen CD1A|T-cell surface antigen T6/Leu-6|cluster of differentiation 1 A|T6|CD1|R4|CD1a molecule|a polypeptide|T-cell surface glycoprotein CD1a|CD1a antigen|differentiation antigen CD1-alpha-3|CD1A antigen|hTa1 thymocyte antigen|HTA1
- Storage Buffer:0.1M BBS (pH 8.2)
- Storage Temperature:Store at 4C. Do not freeze.
- Concentration:0.5 mg/ml
- Immunogen:Fresh dd miniature swine thymocytes
- Purification:Protein A or G purified
- Cat. No.:102161-764
- Supplier no.:NBP1-28221
The CD1 Antibody (76-7-4) from Novus Biologicals is a mouse monoclonal antibody to CD1. This antibody reacts with porcine. The CD1 Antibody (76-7-4) has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Immunocytochemistry.
Type: Primary
Antigen: CD1
Clonality: Monoclonal
Clone: 76-7-4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Porcine