To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cerebral Cavernous Malformation 2
- Antigen Symbol:CCM2
- Clonality:Monoclonal
- Clone:1E9
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:83605
- Antigen Synonyms:C7orf22OSM|MGC74868|cerebral cavernous malformation 2|malcavernin|Cerebral cavernous malformations 2 protein|MGC4067|chromosome 7 open reading frame 22|MGC4607
- Storage Buffer:PBS (pH 7.3), 1.0% BSA and 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human CCM2(NP_113631) produced in HEK293T cell.
- Purification:Immunogen affinity purified
- Cat. No.:102229-006
- Supplier no.:NBP2-01134
Specifications
About this item
The CCM2 Antibody (1E9) from Novus Biologicals is a mouse monoclonal antibody to CCM2. This antibody reacts with human. The CCM2 Antibody (1E9) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: CCM2
Clonality: Monoclonal
Clone: 1E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human