To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Butyrylcholinesterase/BCHE
- Antigen Symbol:Butyrylcholinesterase
- Clonality:Monoclonal
- Clone:10
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Dog
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:590
- Antigen Synonyms:Acylcholine acylhydrolase|E1|butyrylcholinesterase|cholinesterase 1|Choline esterase II|Butyrylcholine esterase|CHE1cholinesterase|EC 3.1.1.8|Pseudocholinesterase
- Storage Buffer:PBS (pH 7.3), 1.0% BSA and 50% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human BCHE (NP_000046) produced in HEK293T cell.
- Purification:Immunogen affinity purified
- Cat. No.:102233-782
- Supplier no.:NBP2-03523
Specifications
About this item
The Butyrylcholinesterase / BCHE Antibody (1E1) from Novus Biologicals is a mouse monoclonal antibody to Butyrylcholinesterase / BCHE. This antibody reacts with human, canine. The Butyrylcholinesterase / BCHE Antibody (1E1) has been validated for the following applications: Western Blot, Flow Cytometry.
Type: Primary
Antigen: Butyrylcholinesterase/BCHE
Clonality: Monoclonal
Clone: 10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Canine