To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Betacellulin/BTC
- Antigen Symbol:BTC
- Clonality:Monoclonal
- Clone:MM0110-7F37
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Form:Lyophilized
- Gene ID:685
- Antigen Synonyms:probetacellulin|betacellulin
- Storage Buffer:A 0.2 um filtered solution in PBS
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:Human Betacellulin
- Purification:Protein G purified
- Cat. No.:102250-840
- Supplier no.:NBP2-12052
Specifications
About this item
The Betacellulin / BTC Antibody (MM0110-7F37) from Novus Biologicals is a mouse monoclonal antibody to Betacellulin / BTC. This antibody reacts with human. The Betacellulin / BTC Antibody (MM0110-7F37) has been validated for the following applications: Western Blot.
Type: Primary
Antigen: Betacellulin/BTC
Clonality: Monoclonal
Clone: MM0110-7F37
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human