To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:beta 2-Microglobulin
- Antigen Symbol:B2M
- Clonality:Monoclonal
- Clone:B2M-01
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:567
- Antigen Synonyms:beta-2-microglobin|beta chain of MHC class I molecules|beta-2-microglobulin
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Store at 4C in the dark.
- Concentration:1.0 mg/ml
- Immunogen:Purified human beta 2 Microglobulin (NM_004048.2).
- Purification:Ammonium sulfate precipitation
- Cat. No.:102124-044
- Supplier no.:NB110-60015
Specifications
About this item
The beta 2-Microglobulin Antibody (B2M-01) [FITC] from Novus Biologicals is a mouse monoclonal antibody to beta 2-Microglobulin. This antibody reacts with human, bovine (negative), canine (negative), chicken (negative), mouse (negative), rabbit (negative). The beta 2-Microglobulin Antibody (B2M-01) [FITC] has been validated for the following applications: Western Blot, Flow Cytometry.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200
Type: Primary
Antigen: beta 2-Microglobulin
Clonality: Monoclonal
Clone: B2M-01
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Bovine (Negative), Canine (Negative), Chicken (Negative), Mouse (Negative), Rabbit (Negative)