To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Agrin
- Antigen Symbol:Agrin
- Clonality:Monoclonal
- Clone:AGR 131
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2 alpha
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:375790
- Antigen Synonyms:agrin|agrin proteoglycan
- Storage Buffer:PBS (pH 7.2) and 50% Glycerol
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Recombinant rat agrin (C-terminal fragment).
- Purification:Protein G purified
- Cat. No.:102121-470
- Supplier no.:NB110-11069
The Agrin Antibody (AGR 131) from Novus Biologicals is a mouse monoclonal antibody to Agrin. This antibody reacts with mouse, rat. The Agrin Antibody (AGR 131) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Microarray.
Microarray
Type: Primary
Antigen: Agrin
Clonality: Monoclonal
Clone: AGR 131
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 Alpha
Reactivity: Mouse, Rat