To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Unc-5 Homolog C
- Antigen Symbol:UNC5H3/UNC5C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 μg
- Environmentally Preferable:
- Epitope:NCTVSEEPTGID
- Gene ID:8633
- Antigen Synonyms:unc-5 homolog 3|Protein unc-5 homolog C|UNC5H3unc5 (C.elegans homolog) c|unc-5 homolog C (C. elegans)|netrin receptor UNC5C|Protein unc-5 homolog 3
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence NCTVSEEPTGID corresponding to internal region according to NP_003719.2.
- Tested Applications:Peptide
- Purification:Immunogen affinity purified
- Cat. No.:102168-754
- Supplier no.:NBP1-37002
Specifications
About this item
The UNC5H3 / UNC5C Antibody from Novus Biologicals is a goat polyclonal antibody to UNC5H3 / UNC5C. This antibody reacts with human, mouse. The UNC5H3 / UNC5C Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.
Type: Primary
Antigen: UNC5H3/UNC5C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: NCTVSEEPTGID
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse