To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Six Transmembrane Epithelial Antigen of the Prostate 4
- Antigen Symbol:STAMP2/STEAP4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:79689
- Antigen Synonyms:STAMP2DKFZp666D049|EC 1.16.1|alpha-induced protein 9metalloreductase STEAP4|TIARPsix transmembrane prostate protein 2|tumor necrosis-alpha-induced adipose-related protein|SixTransMembrane protein of prostate 2|Six-transmembrane epithelial antigen of prostate 4|FLJ23153|EC 1.16.1.-|Tumor necrosis factor|STEAP family member 4|TNFAIP9
- Storage Buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage Temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence CVDNTLTRIRQGWERN corresponding to C-Terminus according to NP_078912.2.
- Tested Applications:, Peptide
- Purification:Immunogen affinity purified
- Cat. No.:102115-500
- Supplier no.:NB100-68162
Specifications
About this item
The STAMP2 / STEAP4 Antibody from Novus Biologicals is a goat polyclonal antibody to STAMP2 / STEAP4. This antibody reacts with human, mouse. The STAMP2 / STEAP4 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation, Peptide ELISA.
Type: Primary
Antigen: STAMP2/STEAP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse